Assay Details
Gene Symbol: Taf4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081092.1 | NP_001074561.1 | 6 - 7 | 1849 | 79 | |
GenBank mRNA | AY038601.1 | - | 6 - 7 | 705 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
