Assay Details
Gene Symbol: Taf4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081092.1 | NP_001074561.1 | 9 - 10 | 2355 | 112 | |
GenBank mRNA | AK136225.1 | - | 2 - 3 | 199 | 112 | |
AY038601.1 | - | 9 - 10 | 1211 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
