Assay Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008960.2 | NP_032986.1 | 3 - 4 | 1081 | 138 | |
| GenBank mRNA | AK076980.1 | - | 3 - 4 | 1039 | 138 | |
| AK088717.1 | - | 3 - 4 | 1060 | 138 | ||
| AK148736.1 | - | 3 - 4 | 916 | 135 | ||
| AK164302.1 | - | 3 - 4 | 138 | |||
| BC021445.1 | - | 3 - 4 | 430 | 5038272 | 138 | |
| U92437.1 | - | 3 - 4 | 1161 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pten
Genomic Map