Assay Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008960.2 | NP_032986.1 | 7 - 8 | 1662 | 110 | |
| GenBank mRNA | AK030750.1 | - | 3 - 4 | 425 | 110 | |
| AK076980.1 | - | 7 - 8 | 1620 | 110 | ||
| AK148736.1 | - | 7 - 8 | 1497 | 110 | ||
| BC021445.1 | - | 7 - 8 | 1011 | 5038272 | 110 | |
| U92437.1 | - | 7 - 8 | 1742 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pten
Genomic Map