Assay Details
| Gene Symbol: Ash2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080793.2 | NP_001074262.1 | 5 - 6 | 502 | 106 | |
| NM_001286207.1 | NP_001273136.1 | 5 - 6 | 475 | 106 | ||
| NM_011791.3 | NP_035921.2 | 5 - 6 | 592 | 106 | ||
| GenBank mRNA | AB020983.2 | - | 5 - 6 | 574 | 106 | |
| AK087934.1 | - | 5 - 6 | 567 | 106 | ||
| AK132471.1 | - | 5 - 6 | 574 | 106 | ||
| AK145576.1 | - | 5 - 6 | 467 | 106 | ||
| AK145790.1 | - | 5 - 6 | 479 | 106 | ||
| AK145967.1 | - | 5 - 6 | 446 | 106 | ||
| AK146254.1 | - | 5 - 6 | 504 | 106 | ||
| AK146599.1 | - | 5 - 6 | 476 | 106 | ||
| AK146756.1 | - | 5 - 6 | 467 | 106 | ||
| AK146938.1 | - | 5 - 6 | 569 | 106 | ||
| AK178767.1 | - | 1 - 2 | 48 | 106 | ||
| AK218371.1 | - | 1 - 2 | 48 | 106 | ||
| BC012957.1 | - | 5 - 6 | 579 | 4211433 | 106 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ash2l
Genomic Map