Assay Details
Gene Symbol: Dap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164533.1 | NP_001158005.1 | 3 - 4 | 570 | 64 | |
NM_022994.3 | NP_075370.2 | 3 - 4 | 570 | 64 | ||
GenBank mRNA | AJ250375.1 | - | 3 - 4 | 314 | 64 | |
AK014280.1 | - | 3 - 4 | 240 | 64 | ||
AK014889.1 | - | 3 - 4 | 240 | 64 | ||
AK032275.1 | - | 3 - 4 | 242 | 64 | ||
AK088960.1 | - | 3 - 4 | 240 | 64 | ||
BC019566.1 | - | 3 - 4 | 570 | 4235517 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
