Assay Details
| Gene Symbol: Muc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013605.2 | NP_038633.1 | 3 - 4 | 1479 | 135 | |
| GenBank mRNA | BC005441.1 | - | 3 - 4 | 1450 | 3601782 | 135 |
| BG079301.2 | - | 2 - 3 | 108 | 135 | ||
| M84683.1 | - | 3 - 4 | 1429 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Muc1
Genomic Map