Assay Details
| Gene Symbol: Msto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144898.2 | NP_659147.2 | 8 - 9 | 847 | 126 | |
| GenBank mRNA | AK134001.1 | - | 304 | 126 | ||
| AK139708.1 | - | 826 | 126 | |||
| AK163280.1 | - | 8 - 9 | 707 | 126 | ||
| AK166236.1 | - | 8 - 9 | 819 | 126 | ||
| AK166834.1 | - | 1 - 2 | 2371 | 126 | ||
| AK167586.1 | - | 8 - 9 | 795 | 126 | ||
| AK170160.1 | - | 8 - 9 | 830 | 126 | ||
| BC008103.1 | - | 8 - 9 | 797 | 3966815 | 126 | |
| BC108354.1 | - | 8 - 9 | 781 | 5035560 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Msto1
Genomic Map