Assay Details
| Gene Symbol: Krtcap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025327.2 | NP_079603.1 | 3 - 4 | 296 | 98 | |
| GenBank mRNA | AK002479.1 | - | 3 - 4 | 297 | 98 | |
| AK003310.1 | - | 3 - 4 | 264 | 98 | ||
| BC031492.1 | - | 3 - 4 | 249 | 3485092 | 98 | |
| BC043030.1 | - | 3 - 4 | 768 | 5693076 | 98 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Krtcap2
Genomic Map