Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300005836.mm.1 |
NM_080466.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005836.mm.1 |
NM_080466.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005836.mm.1 |
TC0300000767.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300005835.mm.1 |
NM_080466.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005835.mm.1 |
NM_080466.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005835.mm.1 |
TC0300000767.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300005837.mm.1 |
NM_080466.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005837.mm.1 |
NM_080466.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005837.mm.1 |
TC0300000767.mm.1 |
MTA TC |