Assay Details
| Gene Symbol: Pklr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099779.1 | NP_001093249.1 | 7 - 8 | 1031 | 81 | |
| NM_013631.2 | NP_038659.2 | 7 - 8 | 1157 | 81 | ||
| GenBank mRNA | AK149508.1 | - | 6 - 7 | 1032 | 81 | |
| AK149524.1 | - | 6 - 7 | 1030 | 81 | ||
| BC152327.1 | - | 7 - 8 | 1129 | 40141144 | 81 | |
| BC152542.1 | - | 7 - 8 | 1128 | 81 | ||
| D63764.1 | - | 7 - 8 | 1157 | 81 | ||
| S79731.1 | - | 7 - 8 | 1157 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pklr
Genomic Map