Assay Details
Gene Symbol: 2210019I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038157.1 | 5 - 6 | 838 | 84 | ||
GenBank mRNA | AK008755.1 | - | 5 - 6 | 838 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | GS homeobox 1 | Gsx1 |
Gsh-1, Gsh1 |
-37.6kb | Anti-sense | Chr. 5 - 147188696 - 147190947 on NCBI Build GRCm38 |
Downstream | pancreatic and duodenal homeobox 1 | Pdx1 |
IDX-1, IPF-1, Ipf1, Mody4, STF-1, pdx-1 |
39.3kb | Anti-sense | Chr. 5 - 147270131 - 147275149 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2295731 | BG072176.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2295731 | AK008755.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2295731 | AU041718.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 62174 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749563 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891125 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847593 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877919 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358969 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650127 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794651 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 967830 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326081 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145614 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641036 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 91954 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123057 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125883 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926917 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668844 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375075 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48692 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111153 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169765 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151367 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31457 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18732 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138596 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2295731 | BG072176.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2295731 | AK008755.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2295731 | AU041718.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05782992 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01246078 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138596 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05782934 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01246067 | - | - |
Whole Mouse Genome G4122A | A_52_P549908 | AK008755.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102683 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05782992 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138596 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04487460 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172506 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04807264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172497 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04636007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172474 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172684 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172641 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172492 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172471 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172685 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172578 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172576 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172566 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04409859 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04694430 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04487465 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172495 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172678 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04807261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172514 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172569 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172493 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04371906 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172524 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172477 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172508 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172574 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172523 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04636006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04371904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04694435 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172486 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172520 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172488 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04752867 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172484 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172686 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172571 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04356175 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172513 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172640 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172533 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04487468 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04356180 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04356186 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172527 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04752870 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172475 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04810249 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04752865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172510 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172530 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24172682 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1453403_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1453403_at | - | - |
Mouse Genome 430 2.0 Array | 1453403_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500033243.mm.1 | TC0500003622.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500033243.mm.1 | NR_038157.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500033243.mm.1 | NR_038157.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500033244.mm.1 | TC0500003622.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500033244.mm.1 | NR_038157.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500033244.mm.1 | NR_038157.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500033245.mm.1 | TC0500003622.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500033245.mm.1 | NR_038157.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500033245.mm.1 | NR_038157.1 | Refseq |
Mouse Expression Set 430 | 1453403_PM_at | - | - |
Mouse Expression Set 430 | 1453403_at | - | - |
Murine Genome U74v2 Set | 166596_at | - | - |
Murine Genome U74v2 Set | 166596_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453403_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453403_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 617173 | - | - |
