Assay Details
| Gene Symbol: Lzts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130525.1 | NP_001123997.1 | 4 - 5 | 1473 | 76 | |
| NM_001130526.1 | NP_001123998.1 | 4 - 5 | 1465 | 76 | ||
| NM_145503.2 | NP_663478.2 | 4 - 5 | 1517 | 76 | ||
| GenBank mRNA | AK081367.1 | - | 4 - 5 | 1392 | 76 | |
| AK129446.1 | - | 3 - 4 | 1382 | 76 | ||
| AK144154.1 | - | 4 - 5 | 1484 | 76 | ||
| AK169936.1 | - | 4 - 5 | 1508 | 76 | ||
| BC014695.1 | - | 3 - 4 | 1366 | 3157493 | 76 | |
| BC092263.1 | - | 4 - 5 | 1497 | 2654420 | 76 | |
More Information
Set Membership: |
|

Lzts2
Genomic Map