Assay Details
Gene Symbol: Tec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113460.2 | NP_001106931.1 | 14 - 15 | 1433 | 101 | |
NM_001113461.2 | NP_001106932.1 | 13 - 14 | 1367 | 101 | ||
NM_001113464.2 | NP_001106935.1 | 13 - 14 | 1346 | 101 | ||
NM_013689.5 | NP_038717.2 | 13 - 14 | 1326 | 101 | ||
GenBank mRNA | AK139329.1 | - | 13 - 14 | 1342 | 101 | |
AK154457.1 | - | 14 - 15 | 1429 | 101 | ||
AK168452.1 | - | 13 - 14 | 1364 | 101 | ||
AK168460.1 | - | 13 - 14 | 1363 | 101 | ||
AK170588.1 | - | 13 - 14 | 1365 | 101 | ||
BC018230.1 | - | 12 - 13 | 1331 | 101 | ||
BC037071.1 | - | 13 - 14 | 1361 | 5029430 | 101 | |
BC062884.1 | - | 9 - 10 | 909 | 5697182 | 101 | |
S53716.1 | - | 13 - 14 | 1357 | 101 | ||
X55663.1 | - | 13 - 14 | 1361 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
