Assay Details
Gene Symbol: Tec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113460.2 | NP_001106931.1 | 3 - 4 | 330 | 103 | |
NM_001113461.2 | NP_001106932.1 | 3 - 4 | 330 | 103 | ||
NM_001113464.2 | NP_001106935.1 | 2 - 3 | 243 | 103 | ||
NM_013689.5 | NP_038717.2 | 3 - 4 | 330 | 103 | ||
GenBank mRNA | AK133418.1 | - | 3 - 4 | 331 | 103 | |
AK139329.1 | - | 2 - 3 | 239 | 103 | ||
AK154457.1 | - | 3 - 4 | 326 | 103 | ||
AK168452.1 | - | 3 - 4 | 326 | 103 | ||
AK168460.1 | - | 3 - 4 | 326 | 103 | ||
AK170588.1 | - | 3 - 4 | 328 | 103 | ||
BC018230.1 | - | 2 - 3 | 278 | 103 | ||
BC037071.1 | - | 3 - 4 | 324 | 5029430 | 103 | |
CJ047828.1 | - | 3 - 4 | 331 | 103 | ||
X55663.1 | - | 3 - 4 | 365 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_013689.5, X55663.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
