Assay Details
Gene Symbol: Tec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113460.2 | NP_001106931.1 | 5 - 6 | 515 | 88 | |
NM_001113461.2 | NP_001106932.1 | 5 - 6 | 515 | 88 | ||
NM_001113464.2 | NP_001106935.1 | 4 - 5 | 428 | 88 | ||
NM_013689.5 | NP_038717.2 | 5 - 6 | 474 | 88 | ||
GenBank mRNA | AK133418.1 | - | 5 - 6 | 475 | 88 | |
AK139329.1 | - | 4 - 5 | 424 | 88 | ||
AK154457.1 | - | 5 - 6 | 511 | 88 | ||
AK168452.1 | - | 5 - 6 | 511 | 88 | ||
AK168460.1 | - | 5 - 6 | 511 | 88 | ||
AK170588.1 | - | 5 - 6 | 513 | 88 | ||
BC018230.1 | - | 4 - 5 | 463 | 88 | ||
BC037071.1 | - | 5 - 6 | 509 | 5029430 | 88 | |
S53716.1 | - | 4 - 5 | 439 | 88 | ||
X55663.1 | - | 5 - 6 | 509 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
