Assay Details
Gene Symbol: 1600014K23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028046.1 | NP_082322.1 | 2 | 786 | 74 | |
GenBank mRNA | AK005459.1 | - | 1 | 744 | 74 | |
BC049727.1 | - | 2 | 777 | 6770312 | 74 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126207 | BC049727.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126207 | NM_028046.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423602 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563097 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893956 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343044 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232563 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301453 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530810 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72585 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326519 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17114 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139970 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173799 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122198 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126207 | BC049727.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126207 | NM_028046.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122198 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173799 | - | - |
Whole Mouse Genome G4122A | A_52_P48433 | BC049727.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P48433 | NM_028046.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P48433 | AK005459.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173799 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505111 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505205 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15036416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14898005 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14922112 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505215 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505108 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14672193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505134 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505164 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505146 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505139 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15036411 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505138 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14697833 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505110 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505121 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505217 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505105 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505194 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15084441 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505214 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505209 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14755850 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505128 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505165 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505147 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505199 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505107 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14898011 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505202 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15008418 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505124 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505141 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505133 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505117 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14755847 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505219 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505103 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14896781 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505143 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15188415 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32505151 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10600687 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600686 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429793_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10600685 | - | - |
Mouse Genome 430 2.0 Array | 1429793_at | - | - |
Mouse Genome 430 2.0 Array | 1429793_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71996_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71996_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00005510.mm.1 | NM_028046.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00005510.mm.1 | NM_028046.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00005510.mm.1 | TC0X00000830.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00005511.mm.1 | NM_028046.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00005511.mm.1 | NM_028046.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00005511.mm.1 | TC0X00000830.mm.1 | MTA TC |
Mouse Expression Set 430 | 1429793_at | - | - |
Mouse Expression Set 430 | 1429793_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429793_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429793_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 635181 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251706 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1251706 | - | - |
