Assay Details
| Gene Symbol: Mfsd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026660.2 | NP_080936.1 | 5 - 6 | 742 | 75 | |
| GenBank mRNA | AK018736.1 | - | 5 - 6 | 755 | 75 | |
| AK089055.1 | - | 5 - 6 | 759 | 75 | ||
| AK152438.1 | - | 5 - 6 | 728 | 75 | ||
| BC023012.1 | - | 5 - 6 | 742 | 5376230 | 75 | |
| BC064775.1 | - | 5 - 6 | 640 | 30023877 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd10
Genomic Map