Assay Details
| Gene Symbol: Mfsd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026660.2 | NP_080936.1 | 9 - 10 | 1237 | 62 | |
| GenBank mRNA | AK003763.1 | - | 4 - 5 | 505 | 62 | |
| AK018736.1 | - | 9 - 10 | 1250 | 62 | ||
| AK089055.1 | - | 9 - 10 | 1254 | 62 | ||
| BC023012.1 | - | 9 - 10 | 1237 | 5376230 | 62 | |
| BC064775.1 | - | 9 - 10 | 1135 | 30023877 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd10
Genomic Map