Assay Details
| Gene Symbol: Rgs12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163512.1 | NP_001156984.1 | 6 - 7 | 892 | 68 | |
| NM_173402.2 | NP_775578.2 | 9 - 10 | 2939 | 68 | ||
| GenBank mRNA | AK004813.2 | - | 7 - 8 | 845 | 68 | |
| AK014569.1 | - | 6 - 7 | 894 | 68 | ||
| AK145890.1 | - | 9 - 10 | 2888 | 68 | ||
| AK161129.1 | - | 6 - 7 | 893 | 68 | ||
| BC040396.1 | - | 9 - 10 | 2936 | 4461061 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rgs12
Genomic Map