Assay Details
| Gene Symbol: Rgs12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163512.1 | NP_001156984.1 | 8 - 9 | 1165 | 66 | |
| NM_173402.2 | NP_775578.2 | 11 - 12 | 3212 | 66 | ||
| GenBank mRNA | AK004813.2 | - | 9 - 10 | 1118 | 66 | |
| AK014569.1 | - | 8 - 9 | 1167 | 66 | ||
| AK145890.1 | - | 11 - 12 | 3161 | 66 | ||
| AK161129.1 | - | 8 - 9 | 1166 | 66 | ||
| BC040396.1 | - | 11 - 12 | 3209 | 4461061 | 66 | |
More Information
Set Membership: |
|

Rgs12
Genomic Map