Assay Details
| Gene Symbol: Acox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030721.2 | NP_109646.2 | 5 - 6 | 639 | 93 | |
| GenBank mRNA | AJ278430.1 | - | 5 - 6 | 672 | 93 | |
| AK028818.1 | - | 5 - 6 | 1060 | 93 | ||
| AK053960.1 | - | 5 - 6 | 636 | 93 | ||
| AK128922.1 | - | 5 - 6 | 599 | 93 | ||
| AK154112.1 | - | 4 - 5 | 730 | 93 | ||
| AK171598.1 | - | 622 | 93 | |||
| AK194193.1 | - | 2 - 3 | 277 | 93 | ||
| BC044725.2 | - | 5 - 6 | 659 | 5374281 | 93 | |
| BC055019.1 | - | 5 - 6 | 639 | 4162223 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acox3
Genomic Map