Assay Details
| Gene Symbol: Acox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030721.2 | NP_109646.2 | 6 - 7 | 725 | 90 | |
| GenBank mRNA | AK028818.1 | - | 6 - 7 | 1146 | 90 | |
| AK053960.1 | - | 6 - 7 | 722 | 90 | ||
| AK128922.1 | - | 6 - 7 | 685 | 90 | ||
| AK154112.1 | - | 5 - 6 | 816 | 90 | ||
| AK171598.1 | - | 708 | 90 | |||
| BC044725.2 | - | 6 - 7 | 745 | 5374281 | 90 | |
| BC055019.1 | - | 6 - 7 | 725 | 4162223 | 90 | |
More Information
Set Membership: |
|

Acox3
Genomic Map