Assay Details
| Gene Symbol: Rfc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027009.2 | NP_081285.1 | 6 - 7 | 766 | 81 | |
| GenBank mRNA | AK042393.1 | - | 6 - 7 | 767 | 81 | |
| AK145126.1 | - | 6 - 7 | 793 | 81 | ||
| AK167047.1 | - | 6 - 7 | 767 | 81 | ||
| AK209135.1 | - | 3 - 4 | 343 | 81 | ||
| BC026795.1 | - | 6 - 7 | 737 | 4015134 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfc3
Genomic Map