Assay Details
| Gene Symbol: Rfc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027009.2 | NP_081285.1 | 7 - 8 | 867 | 126 | |
| GenBank mRNA | AK145126.1 | - | 7 - 8 | 894 | 126 | |
| AK167047.1 | - | 7 - 8 | 868 | 126 | ||
| BC026795.1 | - | 7 - 8 | 838 | 4015134 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfc3
Genomic Map