Assay Details
| Gene Symbol: Plrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016784.3 | NP_058064.2 | 4 - 5 | 77 | ||
| GenBank mRNA | AF044334.1 | - | 3 - 4 | 312 | 77 | |
| AK075993.1 | - | 4 - 5 | 77 | |||
| AK076007.1 | - | 4 - 5 | 77 | |||
| AK077197.1 | - | 4 - 5 | 77 | |||
| AK080068.1 | - | 4 - 5 | 77 | |||
| BC006750.1 | - | 4 - 5 | 3709134 | 77 | ||
| BC052895.1 | - | 4 - 5 | 378 | 77 | ||
More Information
Set Membership: |
|

Plrg1
Genomic Map