Assay Details
Gene Symbol: Poteg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026256.2 | NP_080532.1 | 6 - 7 | 986 | 156 | |
NM_027618.1 | NP_081894.1 | 6 - 7 | 986 | 156 | ||
GenBank mRNA | AK015012.1 | - | 6 - 7 | 987 | 156 | |
AK015165.1 | - | 6 - 7 | 987 | 156 | ||
AK015473.1 | - | 6 - 7 | 987 | 156 | ||
DQ298444.1 | - | 6 - 7 | 987 | 156 | ||
DQ517440.1 | - | 6 - 7 | 987 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
