Assay Details
Gene Symbol: 4930423M02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038183.1 | 3 - 4 | 309 | 86 | ||
GenBank mRNA | AK015191.1 | - | 3 - 4 | 312 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | inositol monophosphatase domain containing 1 | Impad1 |
1110001C20Rik, AA408880, AI451589, AL022796, B230207P20, Jaws, gPAPP |
-845.3kb | Anti-sense | Chr. 4 - 4764342 - 4793306 on NCBI Build GRCm38 |
Downstream |
family with sequence similarity 110, member B |
Fam110b |
1700012H17Rik, 5031405P22, AW049855 |
2.1kb | Sense | Chr. 4 - 5644078 - 5801101 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
