Assay Details
| Gene Symbol: Kcnj15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039056.2 | NP_001034145.1 | 3 - 4 | 370 | 75 | |
| NM_001271689.1 | NP_001258618.1 | 4 - 5 | 501 | 75 | ||
| NM_001271691.1 | NP_001258620.1 | 3 - 4 | 442 | 75 | ||
| NM_001271693.1 | NP_001258622.1 | 3 - 4 | 397 | 75 | ||
| NM_019664.5 | NP_062638.1 | 4 - 5 | 546 | 75 | ||
| GenBank mRNA | AK165436.1 | - | 3 - 4 | 371 | 75 | |
| AW012236.1 | - | 3 - 4 | 308 | 75 | ||
| BC010794.1 | - | 3 - 4 | 236 | 4221516 | 75 | |
| BC057915.1 | - | 4 - 5 | 361 | 5042647 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK165436.1, BC010794.1, BC057915.1, NM_001039056.2, NM_001271689.1, NM_001271691.1, NM_001271693.1, NM_019664.5 transcripts.
Set Membership: |
|

Kcnj15
Genomic Map