Assay Details
Gene Symbol: Ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011809.3 | NP_035939.3 | 8 - 9 | 1313 | 73 | |
GenBank mRNA | AK014602.2 | - | 8 - 9 | 1235 | 73 | |
AK030537.1 | - | 4 - 5 | 615 | 73 | ||
AK038348.1 | - | 8 - 9 | 1300 | 73 | ||
AK128933.1 | - | 9 - 10 | 1187 | 73 | ||
AK143687.1 | - | 8 - 9 | 1265 | 73 | ||
AK145852.1 | - | 8 - 9 | 1149 | 73 | ||
AK150175.1 | - | 8 - 9 | 1242 | 73 | ||
AK151315.1 | - | 8 - 9 | 1264 | 73 | ||
AK152750.1 | - | 8 - 9 | 1267 | 73 | ||
AK153339.1 | - | 8 - 9 | 1265 | 73 | ||
AK153478.1 | - | 8 - 9 | 1265 | 73 | ||
BC005486.1 | - | 8 - 9 | 1234 | 3486021 | 73 | |
BC005504.1 | - | 8 - 9 | 1174 | 3490813 | 73 | |
CT010235.1 | - | 7 - 8 | 1075 | 73 | ||
J04103.1 | - | 8 - 9 | 1212 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
