Assay Details
Gene Symbol: Chuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007700.2 | NP_031726.2 | 19 - 20 | 2193 | 83 | |
GenBank mRNA | AK035326.1 | - | 19 - 20 | 2150 | 83 | |
AK155328.1 | - | 19 - 20 | 2173 | 83 | ||
BC125266.1 | - | 8 - 9 | 792 | 83 | ||
U12473.1 | - | 19 - 20 | 2161 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
