Assay Details
| Gene Symbol: Chuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162410.1 | NP_001155882.1 | 4 - 5 | 466 | 163 | |
| NM_007700.2 | NP_031726.2 | 4 - 5 | 466 | 163 | ||
| GenBank mRNA | AK018671.1 | - | 4 - 5 | 445 | 163 | |
| AK035326.1 | - | 4 - 5 | 423 | 163 | ||
| AK150039.1 | - | 4 - 5 | 445 | 163 | ||
| AK155393.1 | - | 4 - 5 | 467 | 163 | ||
| AK164236.1 | - | 4 - 5 | 467 | 163 | ||
| BC018243.1 | - | 4 - 5 | 458 | 4511275 | 163 | |
| U12473.1 | - | 4 - 5 | 434 | 163 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chuk
Genomic Map