Assay Details
| Gene Symbol: 4632411P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019492.1 | - | 1 - 2 | 1513 | 78 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2267367 | AK019492.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410729 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692593 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 196143 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3099 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158200 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127743 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2267367 | AK019492.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01063053 | - | - |
| Whole Mouse Genome G4122A | A_52_P1067575 | AK019492.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04615815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04693082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04693083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04206037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23567387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04723116 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1430691_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430691_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430691_at | - | - |
| Mouse Expression Set 430 | 1430691_PM_at | - | - |
| Mouse Expression Set 430 | 1430691_at | - | - |
| Murine Genome U74v2 Set | 171307_PM_r_at | - | - |
| Murine Genome U74v2 Set | 171307_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430691_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430691_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 540151 | - | - |

4632411P08Rik
Genomic Map