Assay Details
Gene Symbol: 1700061D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006850.1 | - | 1 - 2 | 77 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 40289 |
Gm40289 | Anti-sense | Chr. 5 - 37971918 - 37987738 on NCBI Build GRCm38 | ||
Downstream | syntaxin 18 | Stx18 |
1810035L21Rik, 4933425D03Rik, AU041562 |
55.0kb | Anti-sense | Chr. 5 - 38038622 - 38137770 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
