Assay Details
| Gene Symbol: Mtg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083328.1 | NP_001076797.1 | 6 - 7 | 675 | 100 | |
| NM_001083329.1 | NP_001076798.1 | 5 - 6 | 581 | 100 | ||
| NM_028422.1 | NP_082698.1 | 6 - 7 | 720 | 100 | ||
| NM_181424.3 | NP_852089.2 | 7 - 8 | 814 | 100 | ||
| GenBank mRNA | AK040689.1 | - | 6 - 7 | 1204 | 100 | |
| BC025144.1 | - | 7 - 8 | 813 | 4483063 | 100 | |
| BC028465.1 | - | 1 - 2 | 116 | 100 | ||
| DT929445.1 | - | 3 - 4 | 298 | 100 | ||
More Information
Set Membership: |
|

Mtg2
Genomic Map