Assay Details
| Gene Symbol: Slco4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148933.1 | NP_683735.1 | 10 - 11 | 2194 | 97 | |
| GenBank mRNA | AK033598.1 | - | 10 - 11 | 2014 | 97 | |
| AK172688.1 | - | 10 - 11 | 1944 | 97 | ||
| BC030719.1 | - | 10 - 11 | 2194 | 4501966 | 97 | |
| BC030720.1 | - | 10 - 11 | 2143 | 4505508 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slco4a1
Genomic Map