Assay Details
| Gene Symbol: Igsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177886.1 | NP_001171357.1 | 3 - 4 | 477 | 87 | |
| NM_001177887.1 | NP_001171358.1 | 3 - 4 | 481 | 87 | ||
| GenBank mRNA | AF537215.1 | - | 3 - 4 | 415 | 87 | |
| AK086973.1 | - | 3 - 4 | 478 | 87 | ||
| BX511978.1 | - | 3 - 4 | 505 | 87 | ||
More Information
Set Membership: |
|

Igsf5
Genomic Map