Assay Details
Gene Symbol: Erg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB073078.1 | - | 6 - 7 | 836 | 67 | |
AK050922.1 | - | 7 - 8 | 937 | 67 | ||
BC145176.1 | - | 5 - 6 | 727 | 9053727 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
