Assay Details
Gene Symbol: Tmprss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015775.2 | NP_056590.2 | 11 - 12 | 1401 | 92 | |
GenBank mRNA | AF113596.1 | - | 11 - 12 | 1271 | 92 | |
AF199362.1 | - | 11 - 12 | 1282 | 92 | ||
AF243500.1 | - | 11 - 12 | 1271 | 92 | ||
AK141350.1 | - | 11 - 12 | 1376 | 92 | ||
AK146047.1 | - | 11 - 12 | 1285 | 92 | ||
BC038393.1 | - | 11 - 12 | 1240 | 2648878 | 92 | |
BC054348.1 | - | 11 - 12 | 1401 | 2648889 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
