Assay Details
Gene Symbol: Kdm5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011419.3 | NP_035549.1 | 22 - 23 | 3602 | 126 | |
GenBank mRNA | AB221606.1 | - | 8 - 9 | 1255 | 126 | |
AF127244.1 | - | 22 - 23 | 3440 | 126 | ||
BC059077.1 | - | 14 - 15 | 2417 | 6826863 | 126 | |
BC060077.1 | - | 3688 | 126 | |||
CA316252.1 | - | 4 - 5 | 126 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0Y00000005.rn
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
