Assay Details
| Gene Symbol: Nek5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177898.4 | NP_808566.2 | 1 - 2 | 66 | 97 | |
| GenBank mRNA | AK032672.1 | - | 1 - 2 | 68 | 97 | |
| AK054168.1 | - | 1 - 2 | 80 | 97 | ||
| BC053516.1 | - | 1 - 2 | 54 | 6311513 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032672.1, AK054168.1, BC053516.1, NM_177898.4 transcripts.
Set Membership: |
|

Nek5
Genomic Map