Assay Details
Gene Symbol: Nek5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177898.4 | NP_808566.2 | 4 - 5 | 388 | 134 | |
GenBank mRNA | AK032672.1 | - | 4 - 5 | 390 | 134 | |
AK054168.1 | - | 4 - 5 | 402 | 134 | ||
AK165108.1 | - | 2 - 3 | 151 | 134 | ||
BC053516.1 | - | 4 - 5 | 376 | 6311513 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
