Assay Details
Gene Symbol: Nek5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177898.4 | NP_808566.2 | 9 - 10 | 884 | 96 | |
GenBank mRNA | AK032672.1 | - | 9 - 10 | 886 | 96 | |
AK054168.1 | - | 9 - 10 | 898 | 96 | ||
BC053516.1 | - | 9 - 10 | 872 | 6311513 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
