Assay Details
| Gene Symbol: Fgfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079908.2 | NP_001073377.1 | 6 - 7 | 1503 | 66 | |
| NM_001079909.2 | NP_001073378.1 | 5 - 6 | 1242 | 66 | ||
| NM_010206.3 | NP_034336.2 | 6 - 7 | 1509 | 66 | ||
| GenBank mRNA | AF176552.1 | - | 4 - 5 | 534 | 66 | |
| AK028354.1 | - | 5 - 6 | 682 | 66 | ||
| AK049704.1 | - | 6 - 7 | 1469 | 66 | ||
| BC010200.1 | - | 6 - 7 | 1402 | 3967874 | 66 | |
| BC033447.1 | - | 4 - 5 | 505 | 4037899 | 66 | |
| M33760.1 | - | 4 - 5 | 526 | 66 | ||
| U22324.1 | - | 5 - 6 | 807 | 66 | ||
| U23445.1 | - | 4 - 5 | 540 | 66 | ||
| X51893.1 | - | 6 - 7 | 864 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgfr1
Genomic Map