Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 38 - 39 | 5012 | 92 | |
| GenBank mRNA | AB008516.1 | - | 37 - 38 | 4889 | 92 | |
| AK148168.1 | - | 1 - 2 | 95 | 92 | ||
| AK162004.1 | - | 3 - 4 | 317 | 92 | ||
| AK164106.1 | - | 6 - 7 | 613 | 92 | ||
| AK164492.1 | - | 6 - 7 | 1531 | 92 | ||
| BC145489.1 | - | 33 - 34 | 4532 | 92 | ||
More Information
Set Membership: |
|

Ttc3
Genomic Map