Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 42 - 43 | 5643 | 136 | |
| GenBank mRNA | AB008516.1 | - | 41 - 42 | 5520 | 136 | |
| AK148168.1 | - | 5 - 6 | 726 | 136 | ||
| AK162004.1 | - | 7 - 8 | 948 | 136 | ||
| AK164106.1 | - | 10 - 11 | 1244 | 136 | ||
| AK164492.1 | - | 10 - 11 | 2162 | 136 | ||
| BC145489.1 | - | 37 - 38 | 5163 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc3
Genomic Map