Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 43 - 44 | 5775 | 70 | |
| GenBank mRNA | AB008516.1 | - | 42 - 43 | 5652 | 70 | |
| AK148168.1 | - | 6 - 7 | 858 | 70 | ||
| AK162004.1 | - | 8 - 9 | 1080 | 70 | ||
| AK164106.1 | - | 11 - 12 | 1376 | 70 | ||
| BC145489.1 | - | 38 - 39 | 5295 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ttc3
Genomic Map