| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600008372.mm.1 |
NM_009441.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008372.mm.1 |
NM_009441.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008372.mm.1 |
TC1600001057.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600008368.mm.1 |
NM_009441.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008368.mm.1 |
NM_009441.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008368.mm.1 |
TC1600001057.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600008369.mm.1 |
NM_009441.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008369.mm.1 |
NM_009441.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008369.mm.1 |
TC1600001057.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600008373.mm.1 |
NM_009441.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008373.mm.1 |
NM_009441.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008373.mm.1 |
TC1600001057.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600008371.mm.1 |
NM_009441.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008371.mm.1 |
NM_009441.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008371.mm.1 |
TC1600001057.mm.1 |
MTA TC |