Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600008184.mm.1 |
NM_028083.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600008184.mm.1 |
NM_028083.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600008184.mm.1 |
TC1600001050.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600008183.mm.1 |
NM_028083.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600008183.mm.1 |
NM_028083.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600008183.mm.1 |
TC1600001050.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600008180.mm.1 |
NM_028083.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600008180.mm.1 |
NM_028083.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600008180.mm.1 |
TC1600001050.mm.1 |
MTA TC |