Assay Details
| Gene Symbol: Chaf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028083.4 | NP_082359.1 | 7 - 8 | 768 | 91 | |
| GenBank mRNA | AK011243.1 | - | 7 - 8 | 769 | 91 | |
| BC013532.1 | - | 6 - 7 | 678 | 4238801 | 91 | |
More Information
Set Membership: |
|

Chaf1b
Genomic Map